siRNA Details
| Gene Symbol: RNASET2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003730.4 | NP_003721.2 | 9 | 988 | ||
| XM_017011397.1 | XP_016866886.1 | 10 | 767 | |||
| XM_017011398.1 | XP_016866887.1 | 8 | 889 | |||
| XM_017011399.1 | XP_016866888.1 | 8 | 903 | |||
| GenBank mRNA | AJ419865.1 | - | 9 | 942 | ||
| AJ419866.1 | - | 9 | 1662 | |||
| AK124363.1 | - | 2 | 2020 | |||
| AK315467.1 | - | 9 | 949 | |||
| BC001660.2 | - | 9 | 914 | 2959583 | ||
| BC001819.2 | - | 9 | 914 | 2959583 | ||
| BC036282.1 | - | 9 | 975 | |||
| BC039713.1 | - | 9 | 917 | 5225327 | ||
| BC051912.1 | - | 9 | 927 | 5227463 | ||
| U85625.2 | - | 9 | 945 | |||

RNASET2
Genomic Map