siRNA Details
| Gene Symbol: IRS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003749.2 | NP_003740.2 | 1 | 4478 | ||
| GenBank mRNA | AF073310.1 | - | 1 | 4480 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRS2| Gene Symbol: IRS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003749.2 | NP_003740.2 | 1 | 4478 | ||
| GenBank mRNA | AF073310.1 | - | 1 | 4480 | ||