siRNA Details
| Gene Symbol: SNX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003794.3 | NP_003785.1 | 1, 2 | 190 | ||
| XM_017007414.1 | XP_016862903.1 | 1, 2 | 182 | |||
| GenBank mRNA | AF065485.1 | - | 1, 2 | 148 | ||
| AK001835.1 | - | 1, 2 | 152 | |||
| BC018762.1 | - | 1, 2 | 166 | 4641014 | ||
| BI669801.1 | - | 1, 2 | 194 | |||
| DA289870.1 | - | 1, 2 | 192 | |||

SNX4
Genomic Map