siRNA Details
| Gene Symbol: URI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252641.1 | NP_001239570.1 | 8, 9 | 1028 | ||
| NM_003796.3 | NP_003787.2 | 8, 9 | 1343 | |||
| NR_045557.1 | 7, 8 | 1308 | ||||
| XM_005259362.2 | XP_005259419.1 | 8, 9 | 1022 | |||
| XM_005259363.4 | XP_005259420.1 | 8, 9 | 1079 | |||
| XM_011527435.1 | XP_011525737.1 | 7, 8 | 905 | |||
| GenBank mRNA | AF091095.1 | - | 9, 10 | 976 | ||
| AK001437.1 | - | 7, 8 | 1026 | |||
| AK226173.1 | - | 8, 9 | 1328 | |||
| AK292170.1 | - | 8, 9 | 1076 | |||
| AK314313.1 | - | 8, 9 | 1065 | |||
| AW954529.1 | - | 2, 3 | 465 | |||
| BC014933.2 | - | 3, 4 | 557 | |||
| BC026184.2 | - | 8, 9 | 1025 | |||
| BC067259.1 | - | 7, 8 | 1032 | 6500727 | ||

URI1
Genomic Map