siRNA Details
| Gene Symbol: RIOK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003831.3 | NP_003822.2 | 13 | 2443 | ||
| XM_011526242.1 | XP_011524544.1 | 12 | 2315 | |||
| XM_011526243.1 | XP_011524545.1 | 13 | 2300 | |||
| GenBank mRNA | AF013591.1 | - | 13 | 1919 | ||
| AK303938.1 | - | 13 | 1931 | |||
| AK304571.1 | - | 8 | 1138 | |||
| BC028360.1 | - | 12 | 2071 | 4828351 | ||

RIOK3
Genomic Map