siRNA Details
| Gene Symbol: FBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003837.3 | NP_003828.2 | 7 | 1187 | ||
| GenBank mRNA | AJ573829.1 | - | 1 | 259 | ||
| Y10812.2 | - | 7 | 1148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBP2| Gene Symbol: FBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003837.3 | NP_003828.2 | 7 | 1187 | ||
| GenBank mRNA | AJ573829.1 | - | 1 | 259 | ||
| Y10812.2 | - | 7 | 1148 | |||