siRNA Details
| Gene Symbol: PEX11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184795.1 | NP_001171724.1 | 4 | 901 | ||
| NM_003846.2 | NP_003837.1 | 4 | 1095 | |||
| NR_073491.1 | 4 | 1091 | ||||
| NR_073492.1 | 4 | 1085 | ||||
| NR_073493.2 | 4 | 1307 | ||||
| GenBank mRNA | AB018080.1 | - | 4 | 877 | ||
| AF093670.1 | - | 4 | 875 | |||
| AK023991.1 | - | 4 | 859 | |||
| AK094173.1 | - | 1 | 1695 | |||
| AK301983.1 | - | 4 | 861 | |||
| BC011963.1 | - | 4 | 875 | 3866690 | ||
| BG435901.1 | - | 1 | 50 | |||

PEX11B
Genomic Map