siRNA Details
| Gene Symbol: AKAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003886.2 | NP_003877.2 | 5 | 2202 | ||
| NM_139289.1 | NP_647450.1 | 5 | 2175 | |||
| GenBank mRNA | AF072756.1 | - | 5 | 2230 | ||
| AK292463.1 | - | 5 | 2196 | |||
| BC033007.1 | - | 5 | 2195 | |||
| BC070150.1 | - | 6 | 1073 | |||
| BC126250.1 | - | 5 | 2201 | 8991966 | ||
| BC126252.1 | - | 5 | 2201 | 8991968 | ||
| Y15195.1 | - | 5 | 2208 | |||

AKAP4
Genomic Map