siRNA Details
| Gene Symbol: EIF3J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284335.1 | NP_001271264.1 | 7 | 1978 | ||
| NM_001284336.1 | NP_001271265.1 | 6 | 1993 | |||
| NM_003758.3 | NP_003749.2 | 8 | 2140 | |||
| GenBank mRNA | AK002006.1 | - | 8 | 2060 | ||
| U97670.1 | - | 9 | 2006 | |||

EIF3J
Genomic Map