siRNA Details
| Gene Symbol: SELENBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258288.1 | NP_001245217.1 | 11 | 1491 | ||
| NM_001258289.1 | NP_001245218.1 | 12 | 1886 | |||
| NM_003944.3 | NP_003935.2 | 12 | 1677 | |||
| GenBank mRNA | AK023875.1 | - | 8 | 2768 | ||
| AK296661.1 | - | 12 | 1855 | |||
| AK298355.1 | - | 12 | 1542 | |||
| AK303815.1 | - | 11 | 1457 | |||
| BC009084.1 | - | 12 | 1631 | 3853674 | ||
| BC032997.1 | - | 10 | 2062 | 4839079 | ||
| EU794660.1 | - | 12 | 1631 | |||

SELENBP1
Genomic Map