siRNA Details
| Gene Symbol: MYOM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003970.3 | NP_003961.3 | 6 | 766 | ||
| XM_006716237.1 | XP_006716300.1 | 6 | 731 | |||
| GenBank mRNA | BC052969.1 | - | 6 | 719 | 6205359 | |
| X69089.1 | - | 6 | 676 | |||

MYOM2
Genomic Map