siRNA Details
| Gene Symbol: SH2D2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161441.1 | NP_001154913.1 | 4 | 558 | ||
| NM_001161442.1 | NP_001154914.1 | 4 | 497 | |||
| NM_001161443.1 | NP_001154915.1 | 3 | 407 | |||
| NM_001161444.1 | NP_001154916.1 | 4 | 528 | |||
| NM_003975.3 | NP_003966.2 | 4 | 528 | |||
| XM_006711615.2 | XP_006711678.1 | 4 | 546 | |||
| XM_017002762.1 | XP_016858251.1 | 4 | 575 | |||
| XM_017002763.1 | XP_016858252.1 | 4 | 566 | |||
| XM_017002764.1 | XP_016858253.1 | 4 | 513 | |||
| XM_017002765.1 | XP_016858254.1 | 4 | 526 | |||
| XM_017002766.1 | XP_016858255.1 | 4 | 516 | |||
| XM_017002767.1 | XP_016858256.1 | 4 | 581 | |||
| GenBank mRNA | AF051325.1 | - | 4 | 450 | ||
| AF097744.1 | - | 4 | 427 | |||
| AJ000553.1 | - | 4 | 474 | |||
| AK222737.1 | - | 4 | 451 | |||
| AK313706.1 | - | 4 | 522 | |||
| AY763098.1 | - | 3 | 407 | |||
| AY763099.1 | - | 4 | 504 | |||
| AY763100.1 | - | 4 | 434 | |||
| BC012107.1 | - | 4 | 490 | 4636736 | ||
| DB128222.1 | - | 4 | 528 | |||

SH2D2A
Genomic Map