siRNA Details
| Gene Symbol: ZNF235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004234.4 | NP_004225.3 | 5 | 2603 | ||
| XM_005259412.4 | XP_005259469.1 | 5 | 6659 | |||
| GenBank mRNA | AK226126.1 | - | 1 | 3632 | ||
| AK300317.1 | - | 5 | 2603 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF235| Gene Symbol: ZNF235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004234.4 | NP_004225.3 | 5 | 2603 | ||
| XM_005259412.4 | XP_005259469.1 | 5 | 6659 | |||
| GenBank mRNA | AK226126.1 | - | 1 | 3632 | ||
| AK300317.1 | - | 5 | 2603 | |||