siRNA Details
| Gene Symbol: TRIP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288962.1 | NP_001275891.1 | 2 | 195 | ||
| NM_001288963.1 | NP_001275892.1 | 2 | 195 | |||
| NM_004240.3 | NP_004231.1 | 2 | 195 | |||
| NR_110231.1 | 2 | 195 | ||||
| XM_005259683.2 | XP_005259740.1 | 2 | 195 | |||
| XM_006722940.1 | XP_006723003.1 | 2 | 195 | |||
| GenBank mRNA | AB007845.1 | - | 2 | 170 | ||
| AB072596.1 | - | 2 | 170 | |||
| AF380114.1 | - | 2 | 115 | |||
| AF502289.1 | - | 2 | 154 | |||
| AJ000414.1 | - | 2 | 154 | |||
| AK223109.1 | - | 2 | 160 | |||
| AK313296.1 | - | 2 | 177 | |||
| AL832186.1 | - | 2 | 281 | |||
| AY081141.1 | - | 2 | 149 | |||
| BC013002.2 | - | 2 | 140 | 3532036 | ||
| BG494851.1 | - | 199 | ||||
| BT006698.1 | - | 2 | 115 | |||
| BT020167.1 | - | 2 | 115 | |||
| BT020171.1 | - | 2 | 115 | |||
| CB997985.1 | - | 2 | 208 | |||
| CR536513.1 | - | 2 | 115 | |||
| DB198268.1 | - | 2 | 197 | |||

TRIP10
Genomic Map