siRNA Details
| Gene Symbol: DDX3Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122665.3 | NP_001116137.1 | 6 | 781 | ||
| NM_001302552.2 | NP_001289481.1 | 5 | 429 | |||
| NM_001324195.1 | NP_001311124.1 | 5 | 514 | |||
| NM_004660.4 | NP_004651.2 | 5 | 514 | |||
| NR_136716.1 | 5 | 514 | ||||
| NR_136717.1 | 5 | 514 | ||||
| NR_136718.1 | 5 | 514 | ||||
| NR_136719.1 | 4 | 384 | ||||
| NR_136720.1 | 5 | 514 | ||||
| NR_136721.1 | 5 | 442 | ||||
| NR_136722.1 | 5 | 429 | ||||
| NR_136723.1 | 5 | 429 | ||||
| NR_136724.1 | 5 | 429 | ||||
| GenBank mRNA | AF000984.1 | - | 5 | 434 | ||
| AF000985.1 | - | 5 | 434 | |||
| AK296366.1 | - | 5 | 418 | |||
| AK302172.1 | - | 5 | 429 | |||
| AK303176.1 | - | 5 | 467 | |||
| AK310382.1 | - | 5 | 419 | |||
| BC034942.2 | - | 6 | 672 | 4826417 | ||
| DC394594.1 | - | 5 | 514 | |||

DDX3Y
Genomic Map