siRNA Details
| Gene Symbol: EIF1AY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278612.1 | NP_001265541.1 | 6 | 817 | ||
| NM_004681.3 | NP_004672.2 | 7 | 868 | |||
| GenBank mRNA | AF000987.1 | - | 7 | 839 | ||
| BC005248.1 | - | 7 | 750 | 3933718 | ||
| DQ786267.1 | - | 6 | 740 | |||

EIF1AY
Genomic Map