siRNA Details
| Gene Symbol: DYRK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004714.2 | NP_004705.1 | 5 | 702 | ||
| NM_006483.2 | NP_006474.1 | 5 | 702 | |||
| NM_006484.2 | NP_006475.1 | 5 | 702 | |||
| XM_005259398.4 | XP_005259455.1 | 5 | 1154 | |||
| GenBank mRNA | AF205861.1 | - | 5 | 599 | ||
| AK074971.1 | - | 5 | 618 | |||
| AK302127.1 | - | 5 | 631 | |||
| BC018751.1 | - | 5 | 654 | 4300012 | ||
| BC025291.1 | - | 5 | 605 | 5019417 | ||
| Y17999.1 | - | 5 | 670 | |||

DYRK1B
Genomic Map