siRNA Details
| Gene Symbol: NOLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284388.1 | NP_001271317.1 | 3 | 438 | ||
| NM_001284389.1 | NP_001271318.1 | 3 | 441 | |||
| NM_004741.4 | NP_004732.2 | 3 | 438 | |||
| XM_005270273.1 | XP_005270330.1 | 3 | 415 | |||
| GenBank mRNA | AK225101.1 | - | 3 | 228 | ||
| AK314363.1 | - | 3 | 225 | |||
| AY820769.1 | - | 3 | 206 | |||
| BC001883.1 | - | 3 | 213 | 3533568 | ||
| D21262.1 | - | 3 | 199 | |||
| DA588285.1 | - | 3 | 438 | |||
| Z34289.1 | - | 3 | 257 | |||

NOLC1
Genomic Map