siRNA Details
| Gene Symbol: CDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003818.3 | NP_003809.1 | 2 | 418 | ||
| XM_006723660.1 | XP_006723723.1 | 2 | 409 | |||
| XM_011529390.1 | XP_011527692.1 | 3 | 741 | |||
| GenBank mRNA | AF069532.1 | - | 1 | 14 | ||
| AK001414.1 | - | 2 | 139 | |||
| AK027695.1 | - | 2 | 125 | |||
| AK091045.1 | - | 3 | 741 | |||
| AK315489.1 | - | 2 | 399 | |||
| BC025751.1 | - | 2 | 139 | 5199748 | ||
| BP353230.1 | - | 2 | 418 | |||
| Y16521.1 | - | 2 | 344 | |||

CDS2
Genomic Map