siRNA Details
| Gene Symbol: NAPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003827.3 | NP_003818.2 | 1, 2 | 426 | ||
| NR_038456.1 | 1, 2 | 426 | ||||
| XM_011527436.1 | XP_011525738.1 | 1, 2 | 100 | |||
| GenBank mRNA | AK098401.1 | - | 1, 2 | 259 | ||
| AK292244.1 | - | 1, 2 | 190 | |||
| AK308633.1 | - | 2, 3 | 316 | |||
| AK310185.1 | - | 1, 2 | 183 | |||
| AY927609.1 | - | 1, 2 | 208 | |||
| BC001165.1 | - | 1, 2 | 198 | 3533668 | ||
| BC007432.2 | - | 1, 2 | 189 | 2962847 | ||
| BC028234.1 | - | 1, 2 | 244 | 5245770 | ||
| BC091511.1 | - | 1, 2 | 205 | 30512911 | ||
| BQ948081.1 | - | 1, 2 | 408 | |||
| BT019568.1 | - | 1, 2 | 108 | |||
| U39412.1 | - | 1, 2 | 175 | |||

NAPA
Genomic Map