siRNA Details
| Gene Symbol: EIF4E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276336.1 | NP_001263265.1 | 2 | 249 | ||
| NM_001276337.1 | NP_001263266.1 | 2 | 249 | |||
| NM_001282958.1 | NP_001269887.1 | 2 | 249 | |||
| NM_004846.3 | NP_004837.1 | 2 | 249 | |||
| XM_005246975.3 | XP_005247032.1 | 2 | 200 | |||
| XM_011512206.2 | XP_011510508.1 | 2 | 524 | |||
| XM_017005377.1 | XP_016860866.1 | 2 | 200 | |||
| GenBank mRNA | AB209519.1 | - | 2 | 153 | ||
| AF038957.1 | - | 2 | 142 | |||
| AF047695.1 | - | 2 | 140 | |||
| AF068117.1 | - | 2 | 125 | |||
| AK022518.1 | - | 2 | 147 | |||
| AK303802.1 | - | 2 | 147 | |||
| BC005392.1 | - | 2 | 147 | 3997986 | ||
| BC005874.2 | - | 2 | 169 | 2961437 | ||
| BC021226.2 | - | 2 | 113 | 4302323 | ||
| BC021690.1 | - | 2 | 151 | 3961350 | ||
| BG913475.1 | - | 2 | 150 | |||
| CB994873.1 | - | 217 | ||||
| CD684451.1 | - | 2 | 172 | |||
| DA992544.1 | - | 2 | 249 | |||

EIF4E2
Genomic Map