siRNA Details
| Gene Symbol: ITM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171581.1 | NP_001165052.1 | 1, 2 | 458 | ||
| NM_004867.4 | NP_004858.1 | 2, 3 | 590 | |||
| GenBank mRNA | AB209310.1 | - | 1 | 4168 | ||
| AF038953.1 | - | 2, 3 | 392 | |||
| AK223327.1 | - | 2, 3 | 369 | |||
| AK303238.1 | - | 1, 2 | 240 | |||
| AK313154.1 | - | 2, 3 | 548 | |||
| AY359051.1 | - | 2, 3 | 331 | |||
| BC040437.1 | - | 2, 3 | 345 | 5748519 | ||
| BG506587.1 | - | 2, 3 | 589 | |||
| CR456875.1 | - | 2, 3 | 253 | |||

ITM2A
Genomic Map