siRNA Details
| Gene Symbol: ITM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171581.1 | NP_001165052.1 | 5 | 1270 | ||
| NM_004867.4 | NP_004858.1 | 6 | 1402 | |||
| GenBank mRNA | AK223327.1 | - | 6 | 1182 | ||
| AK303238.1 | - | 5 | 1053 | |||
| AY359051.1 | - | 6 | 1145 | |||
| BC040437.1 | - | 6 | 1159 | 5748519 | ||
| BM561934.1 | - | 3 | 646 | |||

ITM2A
Genomic Map