siRNA Details
| Gene Symbol: VPS26A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035260.2 | NP_001030337.1 | 8 | 1242 | ||
| NM_001318944.1 | NP_001305873.1 | 10 | 1491 | |||
| NM_001318945.1 | NP_001305874.1 | 10 | 1461 | |||
| NM_001318946.1 | NP_001305875.1 | 8 | 1309 | |||
| NM_004896.4 | NP_004887.2 | 9 | 1385 | |||
| GenBank mRNA | AF054179.1 | - | 9 | 1339 | ||
| AF175266.1 | - | 9 | 1339 | |||
| AK022992.1 | - | 9 | 1315 | |||
| BC022505.1 | - | 9 | 1372 | 4799288 | ||
| BX647725.1 | - | 8 | 1230 | |||
| BX649092.1 | - | 8 | 1285 | |||

VPS26A
Genomic Map