siRNA Details
| Gene Symbol: JMJD7-PLA2G4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198588.1 | NP_001185517.1 | 6 | 692 | ||
| NM_005090.3 | NP_005081.1 | 6 | 692 | |||
| GenBank mRNA | AB209150.1 | - | 6 | 682 | ||
| AF065215.1 | - | 6 | 667 | |||
| AF065216.1 | - | 5 | 743 | |||
| AF121908.1 | - | 6 | 659 | |||
| DQ523799.1 | - | 6 | 660 | |||
| DQ523800.1 | - | 6 | 660 | |||
| Gene Symbol: JMJD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114632.1 | NP_001108104.1 | 6 | 692 | ||
| GenBank mRNA | AK000550.1 | - | 6 | 798 | ||
| AK000663.1 | - | 6 | 666 | |||
| AK000814.1 | - | 653 | ||||
| AK225503.1 | - | 6 | 666 | |||
| AK289833.1 | - | 6 | 692 | |||
| AK290051.1 | - | 6 | 661 | |||
| BC025290.1 | - | 6 | 654 | |||
| BC139899.1 | - | 6 | 671 | |||

JMJD7-PLA2G4B
Genomic Map