siRNA Details
| Gene Symbol: CER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005454.2 | NP_005445.1 | 1, 2 | 556 | ||
| GenBank mRNA | AF400435.1 | - | 1, 2 | 511 | ||
| BC069371.1 | - | 1, 2 | 511 | |||
| BC069405.1 | - | 1, 2 | 511 | 7262136 | ||
| BC069491.1 | - | 1, 2 | 511 | 7262148 | ||
| BC069503.1 | - | 1, 2 | 511 | 7262160 | ||
| BC103976.1 | - | 1, 2 | 511 | 40015380 | ||
| BC103977.1 | - | 1, 2 | 511 | 40015381 | ||
| CD652441.1 | - | 1, 2 | 576 | |||

CER1
Genomic Map