siRNA Details
| Gene Symbol: TROAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005480.3 | NP_005471.3 | 4 | 521 | ||
| XM_006719181.1 | XP_006719244.1 | 4 | 465 | |||
| XM_011537723.1 | XP_011536025.1 | 4 | 465 | |||
| XM_011537724.1 | XP_011536026.1 | 3 | 261 | |||
| GenBank mRNA | AK299302.1 | - | 4 | 426 | ||
| AK309716.1 | - | 3 | 261 | |||
| AK310709.1 | - | 3 | 259 | |||
| AK315211.1 | - | 4 | 465 | |||
| BC032583.1 | - | 4 | 433 | 5498800 | ||
| U04810.2 | - | 4 | 464 | |||

TROAP
Genomic Map