siRNA Details
| Gene Symbol: CHAF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005483.2 | NP_005474.2 | 5 | 1146 | ||
| XM_011527605.2 | XP_011525907.1 | 5 | 1213 | |||
| XM_011527606.2 | XP_011525908.1 | 5 | 1213 | |||
| XM_011527607.2 | XP_011525909.1 | 4 | 1156 | |||
| GenBank mRNA | BC041606.1 | - | 5 | 1071 | ||
| BC052620.1 | - | 5 | 1145 | |||
| BC067093.1 | - | 5 | 1147 | 6339027 | ||
| U20979.1 | - | 5 | 1032 | |||

CHAF1A
Genomic Map