siRNA Details
| Gene Symbol: TAF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201536.1 | NP_001188465.1 | 10 | 1309 | ||
| NM_005681.3 | NP_005672.1 | 10 | 1309 | |||
| NM_139352.2 | NP_647603.1 | 9 | 1139 | |||
| XM_006711612.1 | XP_006711675.1 | 9 | 1358 | |||
| XM_006711613.3 | XP_006711676.1 | 10 | 1322 | |||
| XM_017002758.1 | XP_016858247.1 | 9 | 1358 | |||
| XM_017002759.1 | XP_016858248.1 | 10 | 1278 | |||
| XM_017002760.1 | XP_016858249.1 | 9 | 1086 | |||
| GenBank mRNA | AK001054.1 | - | 9 | 991 | ||
| AK290970.1 | - | 10 | 1254 | |||
| AK307882.1 | - | 3 | 379 | |||
| AK315740.1 | - | 10 | 1293 | |||
| BC013808.1 | - | 10 | 1289 | 3918866 | ||
| L39060.1 | - | 9 | 1124 | |||

TAF1A
Genomic Map