siRNA Details
| Gene Symbol: HHLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145095.1 | NP_001138567.1 | 16 | 1711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HHLA1| Gene Symbol: HHLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145095.1 | NP_001138567.1 | 16 | 1711 | ||