siRNA Details
| Gene Symbol: TSPAN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168412.1 | NP_001161884.1 | 6 | 898 | ||
| NM_005724.5 | NP_005715.1 | 7 | 1090 | |||
| NM_198902.2 | NP_944492.1 | 6 | 1015 | |||
| XM_017021857.1 | XP_016877346.1 | 7 | 699 | |||
| GenBank mRNA | AF054840.1 | - | 7 | 816 | ||
| AF133423.1 | - | 7 | 891 | |||
| AK001305.1 | - | 7 | 936 | |||
| AK001310.1 | - | 7 | 922 | |||
| AK001326.1 | - | 7 | 1033 | |||
| AK027793.1 | - | 4 | 2141 | |||
| AK075336.1 | - | 7 | 971 | |||
| AK075482.1 | - | 6 | 896 | |||
| AK293677.1 | - | 8 | 879 | |||
| AK298157.1 | - | 6 | 779 | |||
| BC000704.2 | - | 7 | 991 | 3349181 | ||
| BC004280.1 | - | 7 | 952 | 3616380 | ||
| BC009248.2 | - | 7 | 955 | 2964568 | ||
| BC011206.1 | - | 7 | 984 | 3457158 | ||
| M69023.1 | - | 3 | 364 | |||

TSPAN3
Genomic Map