siRNA Details
| Gene Symbol: ARL4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282431.1 | NP_001269360.1 | 2 | 1227 | ||
| NM_005737.3 | NP_005728.2 | 1 | 1398 | |||
| GenBank mRNA | AB016811.1 | - | 1 | 906 | ||
| AJ579850.1 | - | 2 | 774 | |||
| AJ579851.1 | - | 1 | 949 | |||
| BC089043.1 | - | 1 | 1402 | 6578402 | ||

ARL4C
Genomic Map