siRNA Details
| Gene Symbol: ZNF256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005773.2 | NP_005764.2 | 3 | 1917 | ||
| XM_017026138.1 | XP_016881627.1 | 2 | 1814 | |||
| GenBank mRNA | AF067165.1 | - | 2 | 1626 | ||
| AK314094.1 | - | 2 | 1758 | |||
| BC001438.1 | - | 3 | 1879 | 3138611 | ||
| BT006853.1 | - | 1 | 1223 | |||

ZNF256
Genomic Map