siRNA Details
| Gene Symbol: MPHOSPH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005791.2 | NP_005782.1 | 10 | 2192 | ||
| GenBank mRNA | AK056862.1 | - | 10 | 2192 | ||
| BC041084.1 | - | 10 | 1842 | |||
| BC126389.1 | - | 10 | 2129 | 8992105 | ||
| X98494.1 | - | 10 | 1815 | |||

MPHOSPH10
Genomic Map