siRNA Details
| Gene Symbol: PRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127708.2 | NP_001121180.2 | 6 | 2528 | ||
| NM_001127709.2 | NP_001121181.2 | 5 | 2372 | |||
| NM_001127710.2 | NP_001121182.2 | 4 | 2249 | |||
| NM_001303232.1 | NP_001290161.1 | 6 | 2522 | |||
| NM_005807.4 | NP_005798.3 | 7 | 2651 | |||
| XM_017000002.1 | XP_016855491.1 | 4 | 2668 | |||
| XM_017000003.1 | XP_016855492.1 | 3 | 2235 | |||
| GenBank mRNA | AK057197.1 | - | 5 | 487 | ||
| AK131434.1 | - | 6 | 1222 | |||
| AK297718.1 | - | 4 | 2366 | |||
| AK301343.1 | - | 8 | 1684 | |||
| AK301487.1 | - | 5 | 2522 | |||
| U70136.1 | - | 6 | 2633 | |||

PRG4
Genomic Map