siRNA Details
| Gene Symbol: PRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127708.2 | NP_001121180.2 | 12 | 4820 | ||
| NM_001127709.2 | NP_001121181.2 | 11 | 4664 | |||
| NM_001127710.2 | NP_001121182.2 | 10 | 4541 | |||
| NM_001303232.1 | NP_001290161.1 | 12 | 4814 | |||
| NM_005807.4 | NP_005798.3 | 13 | 4943 | |||
| GenBank mRNA | U70136.1 | - | 12 | 4925 | ||

PRG4
Genomic Map