siRNA Details
| Gene Symbol: GDF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005811.3 | NP_005802.1 | 3 | 903 | ||
| XM_006719194.3 | XP_006719257.1 | 3 | 905 | |||
| GenBank mRNA | AF028333.1 | - | 3 | 784 | ||
| AF100907.1 | - | 3 | 904 | |||
| KC561773.1 | - | 2 | 525 | |||

GDF11
Genomic Map