siRNA Details
| Gene Symbol: CDC123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006023.2 | NP_006014.2 | 4, 5 | 522 | ||
| GenBank mRNA | AK225634.1 | - | 4, 5 | 488 | ||
| AK289362.1 | - | 4, 5 | 259 | |||
| AK308790.1 | - | 4, 5 | 296 | |||
| BC001600.1 | - | 4, 5 | 272 | 3542611 | ||
| BC009598.1 | - | 4, 5 | 287 | 4134376 | ||
| BP367374.1 | - | 4, 5 | 522 | |||
| CR456966.1 | - | 4, 5 | 238 | |||
| D14878.1 | - | 4, 5 | 518 | |||
| U27112.1 | - | 4, 5 | 300 | |||

CDC123
Genomic Map