siRNA Details
| Gene Symbol: NPEPPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006310.3 | NP_006301.3 | 17 | 2169 | ||
| XM_011525496.1 | XP_011523798.1 | 18 | 1971 | |||
| XM_017025372.1 | XP_016880861.1 | 17 | 2131 | |||
| XM_017025373.1 | XP_016880862.1 | 18 | 1971 | |||
| GenBank mRNA | AJ132583.1 | - | 17 | 1899 | ||
| AK096491.1 | - | 2 | 627 | |||
| AK096709.1 | - | 19 | 1956 | |||
| AK293450.1 | - | 11 | 1101 | |||
| AK293995.1 | - | 10 | 1083 | |||
| AK296887.1 | - | 16 | 1812 | |||
| AK297107.1 | - | 16 | 1673 | |||
| AK303037.1 | - | 18 | 1964 | |||
| BC065294.1 | - | 17 | 1858 | 6059589 | ||

NPEPPS
Genomic Map