siRNA Details
| Gene Symbol: PAGE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318877.1 | NP_001305806.1 | 4 | 334 | ||
| NM_007003.4 | NP_008934.1 | 4 | 346 | |||
| GenBank mRNA | AF275258.1 | - | 4 | 332 | ||
| AJ005894.1 | - | 4 | 330 | |||
| AK312039.1 | - | 4 | 343 | |||
| AL548759.3 | - | 3 | 297 | |||
| BC010897.2 | - | 3 | 296 | 4043535 | ||
| BG435968.1 | - | 4 | 347 | |||
| CR456823.1 | - | 3 | 267 | |||
| CR542183.1 | - | 3 | 267 | |||
| DA841276.1 | - | 4 | 328 | |||

PAGE4
Genomic Map