siRNA Details
| Gene Symbol: CLDN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185072.2 | NP_001172001.1 | 4 | 1886 | ||
| NM_001185073.2 | NP_001172002.1 | 3 | 1847 | |||
| NM_012129.4 | NP_036261.1 | 3 | 1843 | |||
| GenBank mRNA | AL136770.1 | - | 3 | 1695 | ||
| BC036754.2 | - | 2 | 1632 | 5201423 | ||
| BC068532.1 | - | 3 | 1729 | 5294796 | ||

CLDN12
Genomic Map