siRNA Details
| Gene Symbol: CLDN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185072.2 | NP_001172001.1 | 4 | 3488 | ||
| NM_001185073.2 | NP_001172002.1 | 3 | 3449 | |||
| NM_012129.4 | NP_036261.1 | 3 | 3445 | |||
| GenBank mRNA | AL136770.1 | - | 3 | 3297 | ||
| BC036754.2 | - | 2 | 3234 | 5201423 | ||
| BC068532.1 | - | 3 | 3331 | 5294796 | ||

CLDN12
Genomic Map