siRNA Details
| Gene Symbol: PDCD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267556.1 | NP_001254485.1 | 6 | 633 | ||
| NM_001267557.1 | NP_001254486.1 | 4 | 435 | |||
| NM_001267558.1 | NP_001254487.1 | 7 | 554 | |||
| NM_001267559.1 | NP_001254488.1 | 4 | 370 | |||
| NM_013232.3 | NP_037364.1 | 6 | 639 | |||
| GenBank mRNA | AF035606.1 | - | 6 | 665 | ||
| AK001917.1 | - | 6 | 584 | |||
| AK124302.1 | - | 2 | 1894 | |||
| AK128087.1 | - | 3520 | ||||
| AK223366.1 | - | 6 | 639 | |||
| AK315370.1 | - | 6 | 639 | |||
| BC012384.1 | - | 6 | 607 | 3859672 | ||
| BC106706.2 | - | 6 | 588 | 40003290 | ||
| BC110291.1 | - | 6 | 597 | 5187550 | ||
| BG390506.1 | - | 4 | 365 | |||
| BT020072.1 | - | 6 | 539 | |||
| CR983268.1 | - | 4 | 320 | |||
| U58773.1 | - | 6 | 570 | |||

PDCD6
Genomic Map