siRNA Details
| Gene Symbol: TENM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163278.1 | NP_001156750.1 | 2 | 421 | ||
| NM_001163279.1 | NP_001156751.1 | 2 | 421 | |||
| NM_014253.3 | NP_055068.2 | 2 | 421 | |||
| XM_011531230.2 | XP_011529532.1 | 5 | 1062 | |||
| XM_017029208.1 | XP_016884697.1 | 5 | 1062 | |||
| XM_017029209.1 | XP_016884698.1 | 6 | 1117 | |||
| XM_017029210.1 | XP_016884699.1 | 4 | 990 | |||
| XM_017029211.1 | XP_016884700.1 | 5 | 1068 | |||
| XM_017029212.1 | XP_016884701.1 | 5 | 679 | |||
| XM_017029213.1 | XP_016884702.1 | 4 | 991 | |||
| XM_017029214.1 | XP_016884703.1 | 5 | 1062 | |||
| XM_017029215.1 | XP_016884704.1 | 5 | 1061 | |||
| XM_017029216.1 | XP_016884705.1 | 5 | 1061 | |||
| GenBank mRNA | AF100772.1 | - | 2 | 421 | ||
| BC140783.1 | - | 2 | 418 | 9021651 | ||
| BC144543.1 | - | 2 | 418 | 9053070 | ||
| HE578281.1 | - | 2 | 357 | |||

TENM1
Genomic Map