siRNA Details
| Gene Symbol: TTC37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014639.3 | NP_055454.1 | 8 | 821 | ||
| GenBank mRNA | AB002370.1 | - | 8 | 821 | ||
| AK307531.1 | - | 7 | 649 | |||
| BC056893.1 | - | 8 | 794 | 6164578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC37| Gene Symbol: TTC37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014639.3 | NP_055454.1 | 8 | 821 | ||
| GenBank mRNA | AB002370.1 | - | 8 | 821 | ||
| AK307531.1 | - | 7 | 649 | |||
| BC056893.1 | - | 8 | 794 | 6164578 | ||