siRNA Details
Gene Symbol: TTC37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014639.3 | NP_055454.1 | 29 | 3275 | ||
GenBank mRNA | AB002370.1 | - | 29 | 3275 | ||
BC056893.1 | - | 29 | 3248 | 6164578 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TTC37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014639.3 | NP_055454.1 | 29 | 3275 | ||
GenBank mRNA | AB002370.1 | - | 29 | 3275 | ||
BC056893.1 | - | 29 | 3248 | 6164578 |