siRNA Details
| Gene Symbol: FAM53B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014661.3 | NP_055476.3 | 3 | 614 | ||
| GenBank mRNA | AK127343.1 | - | 3 | 808 | ||
| BC031654.1 | - | 3 | 522 | 5174078 | ||
| BX344666.2 | - | 3 | 612 | |||
| CA397342.1 | - | 3 | 527 | |||
| D50930.1 | - | 3 | 303 | |||

FAM53B
Genomic Map