siRNA Details
| Gene Symbol: FAM53B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014661.3 | NP_055476.3 | 4 | 753 | ||
| GenBank mRNA | AK127343.1 | - | 4 | 947 | ||
| BC031654.1 | - | 4 | 661 | 5174078 | ||
| BX344666.2 | - | 4 | 751 | |||
| D50930.1 | - | 4 | 442 | |||

FAM53B
Genomic Map