siRNA Details
Gene Symbol: EMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057571.1 | - | 2 | 2137 | ||
AK222615.1 | - | 9 | 639 | |||
AK290974.1 | - | 9 | 625 | |||
BC020753.1 | - | 9 | 637 | 4809693 | ||
BC021667.1 | - | 9 | 654 | 3682913 | ||
CR457402.1 | - | 9 | 615 | |||
D14659.1 | - | 9 | 621 |

More Information
Other Species Targeted

Panther Classification:
