siRNA Details
| Gene Symbol: KIAA0355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014686.3 | NP_055501.2 | 14 | 4659 | ||
| XM_005259444.1 | XP_005259501.1 | 14 | 4659 | |||
| XM_017027526.1 | XP_016883015.1 | 10 | 2819 | |||
| GenBank mRNA | AB002353.1 | - | 14 | 4637 | ||

KIAA0355
Genomic Map